siRNA Details
Gene Symbol: LOC101927055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120594.1 | 1 | 1731 | |||
GenBank mRNA | AK123298.1 | - | 1 | 1731 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000582038.2 | - | 1 | 1731 | Chr. 2 - 178776926 - 178779963 [+] on Build GRCh38 |
