siRNA Details
Gene Symbol: UNQ6494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024280.1 | 3 | 976 | |||
GenBank mRNA | AK124333.1 | - | 3 | 945 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000617436.1 | - | 945 | Chr. - - [-] on Build |
