siRNA Details
| Gene Symbol: LINC01304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037881.1 | 3 | 1834 | |||
| GenBank mRNA | BC043283.1 | - | 3 | 1481 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000454455.5 | - | 3 | 1872 | Chr. 2 - 3957655 - 3974070 [-] on Build GRCh38 | |

LINC01304
Genomic Map