siRNA Details
| Gene Symbol: MGC27382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027310.2 | 4 | 725 | |||
| GenBank mRNA | BC015860.1 | - | 4 | 706 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413519.1 | - | 4 | 702 | Chr. 1 - 78229622 - 78369464 [+] on Build GRCh38 | |

MGC27382
Genomic Map