siRNA Details
| Gene Symbol: LINC01522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110027.1 | 2 | 197 | |||
| GenBank mRNA | DB066225.1 | - | 2 | 172 | ||
| DB071041.1 | - | 2 | 165 | |||
| DB088024.1 | - | 2 | 126 | |||
| HY050016.1 | - | 2 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01522| Gene Symbol: LINC01522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110027.1 | 2 | 197 | |||
| GenBank mRNA | DB066225.1 | - | 2 | 172 | ||
| DB071041.1 | - | 2 | 165 | |||
| DB088024.1 | - | 2 | 126 | |||
| HY050016.1 | - | 2 | 198 | |||