siRNA Details
| Gene Symbol: LOC102723661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125991.1 | 5 | 757 | |||
| GenBank mRNA | BX095322.1 | - | 5 | 757 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412513.3 | - | 3 | 595 | Chr. 1 - 95996121 - 96022866 [+] on Build GRCh38 | |

LOC102723661
Genomic Map