siRNA Details
| Gene Symbol: LINC01351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110716.1 | 3 | 424 | |||
| NR_110717.1 | 2 | 213 | ||||
| GenBank mRNA | CR936711.1 | - | 3 | 425 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01351| Gene Symbol: LINC01351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110716.1 | 3 | 424 | |||
| NR_110717.1 | 2 | 213 | ||||
| GenBank mRNA | CR936711.1 | - | 3 | 425 | ||