siRNA Details
| Gene Symbol: LOC101928438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109802.1 | 7 | 880 | |||
| GenBank mRNA | AK057451.1 | - | 7 | 880 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427039.1 | - | 7 | 880 | Chr. 9 - 99585786 - 99819889 [-] on Build GRCh38 | |

LOC101928438
Genomic Map