siRNA Details
| Gene Symbol: LOC101928438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109802.1 | 5 | 477 | |||
| GenBank mRNA | AK057451.1 | - | 5 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928438| Gene Symbol: LOC101928438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109802.1 | 5 | 477 | |||
| GenBank mRNA | AK057451.1 | - | 5 | 477 | ||