siRNA Details
| Gene Symbol: LOC105373876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135525.1 | 2 | 251 | |||
| GenBank mRNA | BC031974.1 | - | 2 | 256 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421907.1 | - | 2 | 255 | Chr. 2 - 216866332 - 216867813 [-] on Build GRCh38 | |

LOC105373876
Genomic Map