siRNA Details
| Gene Symbol: LOC105372751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135520.1 | 5 | 667 | |||
| GenBank mRNA | AK124194.1 | - | 5 | 667 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000453784.2 | - | 5 | 667 | Chr. 21 - 24428740 - 24490307 [+] on Build GRCh38 | |

LOC105372751
Genomic Map