siRNA Details
| Gene Symbol: LINC00857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038464.1 | 1 | 456 | |||
| GenBank mRNA | BC009626.1 | - | 1 | 81 | ||
| BC048265.1 | - | 1 | 462 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422847.1 | - | 1 | 456 | Chr. 10 - 80207710 - 80219657 [+] on Build GRCh38 | |

LINC00857
Genomic Map