siRNA Details
| Gene Symbol: KCND3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX098424.1 | - | 2 | 307 | ||
| DB096999.1 | - | 2 | 239 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441125.1 | - | 2 | 307 | Chr. 1 - 111909336 - 111910161 [+] on Build GRCh38 | |

KCND3-AS1
Genomic Map