siRNA Details
| Gene Symbol: ERICH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073397.1 | 3 | 359 | |||
| GenBank mRNA | BC022082.1 | - | 3 | 313 | ||
| BC038783.1 | - | 2 | 187 | |||
| HY026606.1 | - | 3 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH1-AS1| Gene Symbol: ERICH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073397.1 | 3 | 359 | |||
| GenBank mRNA | BC022082.1 | - | 3 | 313 | ||
| BC038783.1 | - | 2 | 187 | |||
| HY026606.1 | - | 3 | 360 | |||