siRNA Details
| Gene Symbol: ERICH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038783.1 | - | 3 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH1-AS1| Gene Symbol: ERICH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038783.1 | - | 3 | 418 | ||