siRNA Details
| Gene Symbol: LOC101928565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125956.1 | 5 | 479 | |||
| GenBank mRNA | AF085892.1 | - | 5 | 479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928565| Gene Symbol: LOC101928565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125956.1 | 5 | 479 | |||
| GenBank mRNA | AF085892.1 | - | 5 | 479 | ||