siRNA Details
| Gene Symbol: LINC01143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126384.1 | 1 | 192 | |||
| GenBank mRNA | BX088774.1 | - | 1 | 192 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449073.2 | - | 1 | 192 | Chr. 2 - 70887871 - 70889959 [+] on Build GRCh38 | |

LINC01143
Genomic Map