siRNA Details
| Gene Symbol: C3orf79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101337.1 | NP_001094807.1 | 1 | 185 | ||
| GenBank mRNA | AF086445.1 | - | 1 | 185 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623038.1 | - | 185 | Chr. - - [-] on Build | ||

C3orf79
Genomic Map