siRNA Details
| Gene Symbol: LINC00211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110011.1 | 3 | 1949 | |||
| NR_110012.1 | 4 | 1771 | ||||
| GenBank mRNA | AK057187.1 | - | 3 | 1636 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413792.5 | - | 3 | 1670 | Chr. 2 - 37826247 - 37829432 [-] on Build GRCh38 | |

LINC00211
Genomic Map