siRNA Details
| Gene Symbol: LINC00211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110011.1 | 1, 2 | 464 | |||
| GenBank mRNA | AK057187.1 | - | 1, 2 | 151 | ||
| HY016653.1 | - | 1, 2 | 464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00211| Gene Symbol: LINC00211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110011.1 | 1, 2 | 464 | |||
| GenBank mRNA | AK057187.1 | - | 1, 2 | 151 | ||
| HY016653.1 | - | 1, 2 | 464 | |||