siRNA Details
| Gene Symbol: NFIA-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104180.1 | 4 | 348 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000442027.5 | - | 4 | 340 | Chr. 1 - 61248945 - 61253510 [-] on Build GRCh38 | |

NFIA-AS1
Genomic Map