siRNA Details
| Gene Symbol: LOC102724434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130921.1 | 4 | 283 | |||
| NR_130922.1 | 4 | 265 | ||||
| NR_130923.1 | 3 | 244 | ||||
| GenBank mRNA | AK097428.1 | - | 3 | 244 | ||
| DB455175.1 | - | 4 | 265 | |||
| DB458471.1 | - | 4 | 270 | |||

LOC102724434
Genomic Map