siRNA Details
| Gene Symbol: LINC01430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109893.1 | 3 | 298 | |||
| GenBank mRNA | BX090567.1 | - | 2 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01430| Gene Symbol: LINC01430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109893.1 | 3 | 298 | |||
| GenBank mRNA | BX090567.1 | - | 2 | 287 | ||