siRNA Details
| Gene Symbol: SIRPG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110090.1 | 3 | 2330 | |||
| GenBank mRNA | AK093519.1 | - | 3 | 2337 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456177.5 | - | 3 | 2330 | Chr. 20 - 1633508 - 1648472 [+] on Build GRCh38 | |

SIRPG-AS1
Genomic Map