siRNA Details
Gene Symbol: NFIA-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110617.1 | 1 | 111 | |||
NR_110618.1 | 1 | 111 | ||||
GenBank mRNA | BC030753.2 | - | 1 | 111 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NFIA-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110617.1 | 1 | 111 | |||
NR_110618.1 | 1 | 111 | ||||
GenBank mRNA | BC030753.2 | - | 1 | 111 |