siRNA Details
| Gene Symbol: NFIA-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110617.1 | 1 | 57 | |||
| NR_110618.1 | 1 | 57 | ||||
| GenBank mRNA | BC030753.2 | - | 1 | 57 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFIA-AS2| Gene Symbol: NFIA-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110617.1 | 1 | 57 | |||
| NR_110618.1 | 1 | 57 | ||||
| GenBank mRNA | BC030753.2 | - | 1 | 57 | ||