siRNA Details
| Gene Symbol: LOC101929011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135069.1 | 2 | 143 | |||
| GenBank mRNA | BX093419.1 | - | 2 | 143 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439483.1 | - | 2 | 85 | Chr. 11 - 116639480 - 116655189 [+] on Build GRCh38 | |

LOC101929011
Genomic Map