siRNA Details
| Gene Symbol: LOC102723927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110592.1 | 1 | 231 | |||
| GenBank mRNA | AK097934.1 | - | 1 | 231 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429456.1 | - | 1 | 231 | Chr. 2 - 241970683 - 241977276 [+] on Build GRCh38 | |

LOC102723927
Genomic Map