siRNA Details
| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 3 | 2206 | |||
| NR_134558.1 | 3 | 1639 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427447.5 | - | 3 | 2168 | Chr. 21 - 33931160 - 33977691 [+] on Build GRCh38 | |

LINC00649
Genomic Map