siRNA Details
| Gene Symbol: LOC105378137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134590.1 | 2 | 398 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424343.1 | - | 2 | 398 | Chr. 6 - 168225279 - 168226517 [-] on Build GRCh38 | |

LOC105378137
Genomic Map