siRNA Details
| Gene Symbol: LOC401312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122075.1 | 5 | 2433 | |||
| GenBank mRNA | BC042871.1 | - | 5 | 2433 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414116.1 | - | 5 | 2433 | Chr. 7 - 22649960 - 22665533 [-] on Build GRCh38 | |

LOC401312
Genomic Map