siRNA Details
| Gene Symbol: ZEB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040248.2 | 4 | 420 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428623.5 | - | 4 | 420 | Chr. 2 - 144519614 - 144520899 [+] on Build GRCh38 | |

ZEB2-AS1
Genomic Map