siRNA Details
Gene Symbol: LINC01625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033919.1 | 4 | 987 | |||
GenBank mRNA | AK123801.1 | - | 4 | 986 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454788.2 | - | 987 | Chr. 6 - 139470164 - 139473055 [-] on Build GRCh38 |
