siRNA Details
| Gene Symbol: LINC01410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121647.1 | 3 | 1657 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424345.5 | - | 3 | 1657 | Chr. 9 - 62801465 - 62813486 [+] on Build GRCh38 | |

LINC01410
Genomic Map