siRNA Details
Gene Symbol: LINC01410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121647.1 | 3 | 972 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000424345.5 | - | 3 | 972 | Chr. 9 - 62801465 - 62813486 [+] on Build GRCh38 |
