siRNA Details
| Gene Symbol: LINC01410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121647.1 | 3 | 412 | |||
| GenBank mRNA | BC039680.1 | - | 3 | 385 | ||
| CR627148.1 | - | 3 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01410| Gene Symbol: LINC01410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121647.1 | 3 | 412 | |||
| GenBank mRNA | BC039680.1 | - | 3 | 385 | ||
| CR627148.1 | - | 3 | 412 | |||