siRNA Details
| Gene Symbol: LINC01589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131244.1 | 1 | 222 | |||
| GenBank mRNA | BC045163.1 | - | 1 | 196 | ||
| BQ188486.1 | - | 1 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01589| Gene Symbol: LINC01589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131244.1 | 1 | 222 | |||
| GenBank mRNA | BC045163.1 | - | 1 | 196 | ||
| BQ188486.1 | - | 1 | 222 | |||