siRNA Details
| Gene Symbol: MID1IP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046706.1 | 1 | 112 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436893.1 | - | 1 | 112 | Chr. X - 38801568 - 38803883 [-] on Build GRCh38 | |

MID1IP1-AS1
Genomic Map