siRNA Details
| Gene Symbol: LOC105376805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135058.1 | 1 | 47 | |||
| NR_135059.1 | 1 | 47 | ||||
| GenBank mRNA | BX115769.1 | - | 1 | 22 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376805| Gene Symbol: LOC105376805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135058.1 | 1 | 47 | |||
| NR_135059.1 | 1 | 47 | ||||
| GenBank mRNA | BX115769.1 | - | 1 | 22 | ||