siRNA Details
Gene Symbol: LOC101928540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125859.1 | 6 | 426 | |||
GenBank mRNA | BX110992.1 | - | 1 | 73 | ||
DB456186.1 | - | 5 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455530.1 | - | 1 | 40 | Chr. 6 - 75454944 - 75458900 [+] on Build GRCh38 |
