siRNA Details
| Gene Symbol: LOC101928540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125859.1 | 9 | 746 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455530.1 | - | 4 | 360 | Chr. 6 - 75454944 - 75458900 [+] on Build GRCh38 | |

LOC101928540
Genomic Map