siRNA Details
| Gene Symbol: FAM50B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010729.1 | XP_016866218.1 | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602854.5 | - | 1 | 45 | Chr. 6 - 3831933 - 3833896 [+] on Build GRCh38 | |

FAM50B
Genomic Map