siRNA Details
| Gene Symbol: CD81-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108080.1 | 3 | 1072 | |||
| GenBank mRNA | BC019904.1 | - | 3 | 1362 | ||
| BC042822.1 | - | 3 | 1068 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD81-AS1| Gene Symbol: CD81-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108080.1 | 3 | 1072 | |||
| GenBank mRNA | BC019904.1 | - | 3 | 1362 | ||
| BC042822.1 | - | 3 | 1068 | |||