siRNA Details
| Gene Symbol: LOC101927641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110270.1 | 5, 6 | 2031 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439734.5 | - | 4, 5 | 1972 | Chr. 2 - 199624531 - 199659132 [+] on Build GRCh38 | |

LOC101927641
Genomic Map