siRNA Details
| Gene Symbol: LINC00317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038872.1 | 2 | 106 | |||
| GenBank mRNA | BF026848.1 | - | 2 | 87 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419069.1 | - | 2 | 106 | Chr. 21 - 21723293 - 21737319 [-] on Build GRCh38 | |

LINC00317
Genomic Map