siRNA Details
| Gene Symbol: LINC00707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038291.1 | 5 | 2251 | |||
| GenBank mRNA | AK094154.1 | - | 5 | 2257 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436383.1 | - | 5 | 2251 | Chr. 10 - 6779598 - 6842906 [+] on Build GRCh38 | |

LINC00707
Genomic Map