siRNA Details
| Gene Symbol: ATP6V1B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110273.1 | 1 | 104 | |||
| NR_110274.1 | 1 | 104 | ||||
| GenBank mRNA | BM014398.1 | - | 1 | 40 | ||
| BM464091.1 | - | 1 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1B1-AS1| Gene Symbol: ATP6V1B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110273.1 | 1 | 104 | |||
| NR_110274.1 | 1 | 104 | ||||
| GenBank mRNA | BM014398.1 | - | 1 | 40 | ||
| BM464091.1 | - | 1 | 64 | |||