siRNA Details
| Gene Symbol: ST3GAL6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046683.1 | 4 | 588 | |||
| GenBank mRNA | BG821580.1 | - | 4 | 584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ST3GAL6-AS1| Gene Symbol: ST3GAL6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046683.1 | 4 | 588 | |||
| GenBank mRNA | BG821580.1 | - | 4 | 584 | ||