siRNA Details
| Gene Symbol: LINC01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109987.1 | 3 | 354 | |||
| GenBank mRNA | BC038725.2 | - | 2 | 134 | ||
| BI460052.1 | - | 3 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01210| Gene Symbol: LINC01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109987.1 | 3 | 354 | |||
| GenBank mRNA | BC038725.2 | - | 2 | 134 | ||
| BI460052.1 | - | 3 | 169 | |||