siRNA Details
Gene Symbol: LINC01210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109987.1 | 4 | 425 | |||
GenBank mRNA | BC038725.2 | - | 3 | 205 | ||
BI460052.1 | - | 4 | 240 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109987.1 | 4 | 425 | |||
GenBank mRNA | BC038725.2 | - | 3 | 205 | ||
BI460052.1 | - | 4 | 240 |